Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342505.1 | NP_001328584.1 | 8 - 9 | 1022 | 73 | |
NM_001342506.1 | NP_001328583.1 | 9 - 10 | 869 | 73 | ||
NM_120018.4 | NP_195569.1 | 9 - 10 | 825 | 73 | ||
GenBank mRNA | AY086801.1 | - | 9 - 10 | 725 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |