Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160820.1 | NP_001154292.1 | 16 - 17 | 2391 | 82 | |
NM_120020.4 | NP_195571.3 | 16 - 17 | 2455 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |