Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9B16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342521.1 | NP_001329141.1 | 3 - 4 | 868 | 103 | |
NM_120059.3 | NP_195610.3 | 4 - 5 | 793 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |