Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GH9B17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120060.2 | NP_195611.1 | 2 - 3 | 753 | 81 | |
GenBank mRNA | DQ446906.1 | - | 2 - 3 | 710 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |