Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120063.6 | NP_195614.2 | 1 - 2 | 718 | 63 | |
GenBank mRNA | AF416569.1 | - | 1 - 2 | 415 | 63 | |
AK175482.1 | - | 1 - 2 | 488 | 63 | ||
AK176074.1 | - | 1 - 2 | 487 | 63 | ||
AK176272.1 | - | 1 - 2 | 486 | 63 | ||
AK176334.1 | - | 1 - 2 | 489 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |