Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120063.6 | NP_195614.2 | 3 - 4 | 914 | 89 | |
GenBank mRNA | AF416569.1 | - | 3 - 4 | 611 | 89 | |
AK175482.1 | - | 3 - 4 | 684 | 89 | ||
AK176074.1 | - | 3 - 4 | 683 | 89 | ||
AK176272.1 | - | 3 - 4 | 682 | 89 | ||
AK176334.1 | - | 3 - 4 | 685 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |