Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001204030.2 | NP_001190959.1 | 1 - 2 | 265 | 80 | |
NM_120070.3 | NP_568053.1 | 1 - 2 | 265 | 80 | ||
GenBank mRNA | AF277453.1 | - | 1 - 2 | 132 | 80 | |
AY050934.1 | - | 1 - 2 | 114 | 80 | ||
AY079396.1 | - | 1 - 2 | 75 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |