Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IMPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342523.1 | NP_001320164.1 | 8 - 9 | 970 | 67 | |
NM_120072.5 | NP_195623.3 | 8 - 9 | 1057 | 67 | ||
GenBank mRNA | AK175533.1 | - | 8 - 9 | 998 | 67 | |
AK220687.1 | - | 8 - 9 | 998 | 67 | ||
BT010891.1 | - | 8 - 9 | 970 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |