Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342527.1 | NP_001328715.1 | 7 - 8 | 789 | 100 | |
NM_120081.2 | NP_195632.1 | 7 - 8 | 967 | 100 | ||
GenBank mRNA | AY059862.1 | - | 7 - 8 | 907 | 100 | |
BT010378.1 | - | 6 - 7 | 748 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |