Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CESA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120095.4 | NP_195645.1 | 3 - 4 | 1262 | 74 | |
GenBank mRNA | AF027173.2 | - | 3 - 4 | 742 | 74 | |
KJ139038.1 | - | 3 - 4 | 572 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |