Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127025.3 | NP_179068.1 | 1 | 340 | 66 | |
GenBank mRNA | AY064023.1 | - | 1 | 262 | 66 | |
AY117187.1 | - | 1 | 228 | 66 | ||
M90508.1 | - | 1 | 240 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Both primers and probe map within a single exon |