Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001161045.2 | NP_001154517.1 | 9 - 10 | 860 | 139 | |
NM_001202597.1 | NP_001189526.1 | 9 - 10 | 860 | 139 | ||
NM_001335430.1 | NP_001324683.1 | 9 - 10 | 860 | 139 | ||
NM_001335431.1 | NP_001324684.1 | 9 - 10 | 860 | 139 | ||
NM_001335432.1 | NP_001324685.1 | 8 - 9 | 990 | 139 | ||
NM_001335433.1 | NP_001324686.1 | 8 - 9 | 948 | 139 | ||
NM_001335434.1 | NP_001324682.1 | 3 - 4 | 341 | 139 | ||
NM_127032.2 | NP_179075.1 | 8 - 9 | 990 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |