Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127084.3 | NP_179126.2 | 8 - 9 | 1046 | 80 | |
GenBank mRNA | AF530477.1 | - | 8 - 9 | 887 | 80 | |
AK229578.1 | - | 8 - 9 | 945 | 80 | ||
BT025547.1 | - | 8 - 9 | 860 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |