Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127099.3 | NP_179141.1 | 1 | 1447 | 88 | |
NM_201739.3 | NP_973468.2 | 2 | 1655 | 88 | ||
GenBank mRNA | AF417474.1 | - | 1 | 1447 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |