Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THY-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335472.1 | NP_001324592.1 | 6 - 7 | 1396 | 61 | |
NM_001335473.1 | NP_001324593.1 | 5 - 6 | 1141 | 61 | ||
NM_127191.4 | NP_179230.1 | 6 - 7 | 1396 | 61 | ||
GenBank mRNA | AY063968.1 | - | 6 - 7 | 1088 | 61 | |
AY114032.1 | - | 5 - 6 | 979 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |