Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBC23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335502.1 | NP_001324377.1 | 4 - 5 | 2748 | 69 | |
NM_127245.3 | NP_179284.1 | 4 - 5 | 2737 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |