Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036282.2 | NP_001031359.1 | 10 - 11 | 1177 | 150 | |
NM_127248.4 | NP_179287.3 | 10 - 11 | 1115 | 150 | ||
GenBank mRNA | BT002041.1 | - | 10 - 11 | 1103 | 150 | |
BT008804.1 | - | 10 - 11 | 1011 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |