Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036282.2 | NP_001031359.1 | 1 - 2 | 349 | 99 | |
NM_127248.4 | NP_179287.3 | 1 - 2 | 275 | 99 | ||
GenBank mRNA | BT002041.1 | - | 1 - 2 | 275 | 99 | |
BT008804.1 | - | 1 - 2 | 183 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Arabidopsis thaliana Probe spans exons |