Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036282.2 | NP_001031359.1 | 28 - 29 | 2767 | 80 | |
NM_127248.4 | NP_179287.3 | 28 - 29 | 2705 | 80 | ||
GenBank mRNA | BT002041.1 | - | 28 - 29 | 2693 | 80 | |
BT008804.1 | - | 28 - 29 | 2601 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |