Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEE15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335508.1 | NP_001318235.1 | 13 - 14 | 1322 | 89 | |
NM_001335509.1 | NP_001323497.1 | 13 - 14 | 1388 | 89 | ||
NM_001335512.1 | NP_001323502.1 | 11 - 12 | 1067 | 89 | ||
NM_001335514.1 | NP_001323501.1 | 12 - 13 | 1490 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |