Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127264.2 | NP_179301.1 | 4 - 5 | 636 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |