Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127292.3 | NP_179329.1 | 1 - 2 | 1214 | 135 | |
GenBank mRNA | AY057680.1 | - | 1 - 2 | 1141 | 135 | |
AY143930.1 | - | 1 - 2 | 909 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |