Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKR2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127294.5 | NP_179331.1 | 5 - 6 | 710 | 66 | |
GenBank mRNA | BT024730.1 | - | 4 - 5 | 549 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |