Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VPS35A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127333.5 | NP_179370.2 | 20 - 21 | 2276 | 110 | |
GenBank mRNA | BT008553.1 | - | 20 - 21 | 2154 | 110 | |
BT008664.1 | - | 20 - 21 | 2204 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |