Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127335.2 | NP_565424.1 | 11 - 12 | 2831 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |