Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERD7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335562.1 | NP_001323840.1 | 2 - 3 | 1177 | 75 | |
NM_127337.3 | NP_179374.1 | 2 - 3 | 1231 | 75 | ||
GenBank mRNA | AF325067.1 | - | 2 - 3 | 1100 | 75 | |
AF428331.1 | - | 2 - 3 | 1150 | 75 | ||
AY081319.1 | - | 2 - 3 | 1154 | 75 | ||
BT001230.1 | - | 2 - 3 | 1100 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |