Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPDHC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036448.2 | NP_001031525.1 | 4 - 5 | 1143 | 85 | |
NM_001336924.1 | NP_001325243.1 | 2 - 3 | 1837 | 85 | ||
NM_129717.5 | NP_181685.1 | 4 - 5 | 1380 | 85 | ||
NM_180021.5 | NP_850352.1 | 3 - 4 | 1449 | 85 | ||
GenBank mRNA | AJ347019.1 | - | 4 - 5 | 1037 | 85 | |
AY063920.1 | - | 3 - 4 | 1291 | 85 | ||
AY091247.1 | - | 2 - 3 | 882 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |