Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_129782.3 | NP_181749.1 | 1 - 2 | 792 | 65 | |
GenBank mRNA | AJ011638.1 | - | 1 - 2 | 392 | 65 | |
AY150378.1 | - | 1 - 2 | 360 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons |