Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337082.1 | NP_001324514.1 | 11 - 12 | 859 | 142 | |
NM_001337083.1 | NP_001324515.1 | 11 - 12 | 767 | 142 | ||
NM_129998.3 | NP_566016.1 | 11 - 12 | 808 | 142 | ||
NM_180084.2 | NP_850415.1 | 11 - 12 | 811 | 142 | ||
GenBank mRNA | AF387018.1 | - | 11 - 12 | 810 | 142 | |
BT006613.1 | - | 10 - 11 | 676 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |