Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DL1D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180089.2 | NP_850420.1 | 4 - 5 | 550 | 152 | |
GenBank mRNA | AJ304843.1 | - | 4 - 5 | 542 | 152 | |
BT046114.1 | - | 4 - 5 | 408 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |