Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG8E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130080.6 | NP_182042.2 | 4 - 5 | 276 | 91 | |
NM_180100.7 | NP_850431.2 | 3 - 4 | 344 | 91 | ||
GenBank mRNA | AF412106.1 | - | 4 - 5 | 200 | 91 | |
AY074388.1 | - | 4 - 5 | 260 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |