Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337134.1 | NP_001324014.1 | 3 - 4 | 1120 | 82 | |
NM_180107.3 | NP_850438.2 | 4 - 5 | 1113 | 82 | ||
GenBank mRNA | AB493593.1 | - | 3 - 4 | 883 | 82 | |
AY074647.1 | - | 3 - 4 | 887 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |