Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035552.2 | NP_001030629.1 | 5 - 6 | 1465 | 144 | |
NM_111266.2 | NP_566218.1 | 5 - 6 | 1197 | 144 | ||
NM_180179.3 | NP_850510.1 | 5 - 6 | 1453 | 144 | ||
GenBank mRNA | AF419591.1 | - | 5 - 6 | 1196 | 144 | |
AK317103.1 | - | 5 - 6 | 1352 | 144 | ||
AY097342.1 | - | 5 - 6 | 1097 | 144 | ||
AY894117.1 | - | 5 - 6 | 1249 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |