Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337562.1 | NP_001325515.1 | 16 - 17 | 2268 | 64 | |
NM_111381.3 | NP_566246.1 | 16 - 17 | 2385 | 64 | ||
NM_180185.3 | NP_850516.1 | 16 - 17 | 2383 | 64 | ||
GenBank mRNA | AK226843.1 | - | 16 - 17 | 2343 | 64 | |
AY056292.1 | - | 16 - 17 | 2293 | 64 | ||
AY127022.1 | - | 16 - 17 | 2336 | 64 | ||
AY142674.1 | - | 15 - 16 | 1984 | 64 | ||
BT000631.1 | - | 15 - 16 | 1984 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |