Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYP43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111445.5 | NP_566256.1 | 6 - 7 | 1122 | 82 | |
NM_180188.3 | NP_850519.1 | 6 - 7 | 1068 | 82 | ||
GenBank mRNA | AJ245408.1 | - | 6 - 7 | 1044 | 82 | |
AY099723.1 | - | 6 - 7 | 1068 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |