Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATSK12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111458.5 | NP_187235.1 | 2 - 3 | 592 | 80 | |
NM_180189.4 | NP_850520.1 | 2 - 3 | 320 | 80 | ||
GenBank mRNA | AK227034.1 | - | 2 - 3 | 360 | 80 | |
AK316673.1 | - | 2 - 3 | 389 | 80 | ||
AY062713.1 | - | 2 - 3 | 305 | 80 | ||
AY093347.1 | - | 1 - 2 | 114 | 80 | ||
X75431.1 | - | 287 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |