Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111573.5 | NP_187349.2 | 1 - 2 | 330 | 71 | |
NM_180197.2 | NP_850528.1 | 1 - 2 | 283 | 71 | ||
GenBank mRNA | BT015414.1 | - | 1 - 2 | 192 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |