Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ECT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338026.1 | NP_001327857.1 | 6 - 7 | 1172 | 88 | |
NM_001338027.1 | NP_001327858.1 | 5 - 6 | 1438 | 88 | ||
NM_001338028.1 | NP_001327859.1 | 5 - 6 | 1057 | 88 | ||
NM_001338029.1 | NP_001327860.1 | 5 - 6 | 1201 | 88 | ||
NM_112143.4 | NP_187912.2 | 5 - 6 | 1077 | 88 | ||
NM_180241.2 | NP_850572.1 | 5 - 6 | 1057 | 88 | ||
GenBank mRNA | AK228498.1 | - | 5 - 6 | 857 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |