Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VCS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338042.1 | NP_001327264.1 | 2 - 3 | 1025 | 84 | |
NM_112175.3 | NP_187938.2 | 2 - 3 | 1022 | 84 | ||
NM_180245.2 | NP_850576.1 | 3 - 4 | 917 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |