Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KINESIN-13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_112536.3 | NP_188285.1 | 11 - 12 | 2090 | 100 | |
NM_180267.2 | NP_850598.1 | 11 - 12 | 2068 | 100 | ||
GenBank mRNA | AK226569.1 | - | 11 - 12 | 1945 | 100 | |
AY056129.1 | - | 11 - 12 | 1926 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |