Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180274.3 | NP_850605.1 | 1 - 2 | 193 | 74 | |
GenBank mRNA | AF453477.1 | - | 1 - 2 | 110 | 74 | |
AK175440.1 | - | 1 - 2 | 151 | 74 | ||
AK175523.1 | - | 1 - 2 | 102 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |