Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113012.4 | NP_566676.1 | 7 - 8 | 1027 | 90 | |
NM_180287.5 | NP_850618.1 | 7 - 8 | 1031 | 90 | ||
GenBank mRNA | AB119060.1 | - | 7 - 8 | 909 | 90 | |
AY042817.1 | - | 7 - 8 | 998 | 90 | ||
AY064628.1 | - | 6 - 7 | 825 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |