Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DXPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113045.3 | NP_566686.2 | 8 - 9 | 1914 | 75 | |
NM_180289.4 | NP_850620.2 | 8 - 9 | 2015 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |