Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSH7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338685.1 | NP_001326470.1 | 15 - 16 | 2899 | 66 | |
NM_180299.1 | NP_850630.1 | 15 - 16 | 2780 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |