Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120114.2 | NP_195664.2 | 3 - 4 | 537 | 161 | |
NM_202986.2 | NP_974715.1 | 4 - 5 | 403 | 161 | ||
GenBank mRNA | AK117791.1 | - | 3 - 4 | 537 | 161 | |
BT005291.1 | - | 3 - 4 | 310 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |