Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120154.5 | NP_568074.1 | 5 - 6 | 956 | 83 | |
GenBank mRNA | AY058889.1 | - | 5 - 6 | 842 | 83 | |
AY079044.1 | - | 5 - 6 | 713 | 83 | ||
U76845.1 | - | 5 - 6 | 885 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |