Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203270.1 | NP_001190199.1 | 5 - 6 | 1354 | 111 | |
NM_120205.5 | NP_195747.2 | 5 - 6 | 1367 | 111 | ||
GenBank mRNA | AY557187.1 | - | 5 - 6 | 1186 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |