Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ESP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342587.1 | NP_001330013.1 | 3 - 4 | 804 | 120 | |
NM_001342588.1 | NP_001330014.1 | 4 - 5 | 761 | 120 | ||
NM_120218.2 | NP_195760.1 | 4 - 5 | 531 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |