Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCAR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342607.1 | NP_001318456.1 | 4 - 5 | 680 | 55 | |
NM_001342608.1 | NP_001330632.1 | 2 - 3 | 616 | 55 | ||
NM_001342609.1 | NP_001330631.1 | 3 - 4 | 562 | 55 | ||
NM_001342610.1 | NP_001330637.1 | 3 - 4 | 771 | 55 | ||
NM_001342611.1 | NP_001330636.1 | 2 - 3 | 616 | 55 | ||
NM_001342612.1 | NP_001330633.1 | 1 - 2 | 173 | 55 | ||
NM_001342614.1 | NP_001330634.1 | 3 - 4 | 771 | 55 | ||
NM_001342615.1 | NP_001330639.1 | 2 - 3 | 616 | 55 | ||
NM_001342616.1 | NP_001330638.1 | 3 - 4 | 521 | 55 | ||
NM_120251.4 | NP_195793.3 | 3 - 4 | 562 | 55 | ||
GenBank mRNA | AY743927.1 | - | 3 - 4 | 441 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |