Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAPTOR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120255.3 | NP_195797.3 | 18 - 19 | 3477 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |